Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX8 Antibody (OTI1E5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274266PCP
Description
SNX8 Monoclonal specifically detects SNX8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| SNX8 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| SNX8 | |
| Full length recombinant protein of human SNX8 (NP_037453) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1E5 | |
| Flow Cytometry | |
| DKFZp761E1721, Mvp1, sorting nexin 8, sorting nexin-8 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 29886.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction