Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX8 Monoclonal Antibody (OTI4C10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502135S


Description
SNX8 Monoclonal antibody specifically detects SNX8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
SNX8 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y5X2 | |
Mouse | |
Affinity chromatography | |
RUO | |
29886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI4C10 | |
Unconjugated | |
Snx8 | |
B130023O14Rik; Mvp1; SNX8; sorting nexin 8; sorting nexin-8 | |
Full length human recombinant protein of human SNX8 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction