Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563428
Description
Immunogen sequence: EKGVLHKHQR ALHKYSLMKR QMMSATAQNR EPESVEQLES RIVEQENAIQ TMELRNYFSL YCLHQETQLI HVYLPLTSHI LRAFVNSQIQ GHKEMSKVWN DLRPKLSCLF AGPHS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX8 is an ortholog of a yeast protein.
Specifications
SNX8 | |
Polyclonal | |
Unconjugated | |
Snx8 | |
B130023O14Rik; Mvp1; SNX8; sorting nexin 8; sorting nexin-8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29886 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5X2 | |
Snx8 | |
Recombinant protein corresponding to Human SNX8. Recombinant protein control fragment (Product #RP-92668). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction