Learn More
SNX9 Mouse anti-Canine, Human, Clone: OTI2B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501324

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein.Specifications
SNX9 | |
Monoclonal | |
Unconjugated | |
SNX9 | |
SDP1, SH3PX1, SH3PXD3A, WISP | |
Mouse | |
Affinity Chromatography | |
RUO | |
476254, 51429 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5X1 | |
SNX9 | |
Full length human recombit protein of human SNX9 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.