Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102325 Supplier Invitrogen™ Supplier No. PA5102325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SNX9.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Snx9
Gene Accession No. Q91VH2, Q9Y5X1
Gene Alias 2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP
Gene Symbols Snx9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SNX9(Accession Q9Y5X1), corresponding to amino acid residues G429-A479.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51429, 66616, 683687
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.