Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOAT 2 Antibody (OTI2D3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274271B
Description
SOAT 2 Monoclonal specifically detects SOAT 2 in Human samples. It is validated for Immunohistochemistry.Specifications
SOAT 2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SOAT2 | |
Recombinant protein fragment corresponding to amino acids 1-120 of human SOAT 2 (NP_003569) produced in E.coli. | |
0.1 mL | |
8435.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI2D3 | |
Immunohistochemistry | |
ACACT2MGC116732, ACAT-2, ACAT2ARGP2, acyl Co-A: cholesterol acyltransferase 2, acyl coenzyme A:cholesterol acyltransferase 2, acyl-CoA:cholesterol acyltransferase 2, Acyl-coenzyme A:cholesterol acyltransferase 2, Cholesterol acyltransferase 2, EC 2.3.1.26, sterol O-acyltransferase 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction