Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561455 Supplier Invitrogen™ Supplier No. PA561455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVGEEKMSLR NRLSKSRENP EEDEDQRNPA KESLETPSNG RIDIKQLIAK KIKLTAEAEE LKPF Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 67%.

SOAT1 (sterol O-acyltransferase-1), also designated ACAT1, is a homotetra-meric enzyme that catalyzes the formation of cholesterol esters from cholesterol and long chain fatty acyl-coenzyme A (acyl-CoA). The gene encoding human SOAT1 maps to chromosome 1 and is expressed as a protein that localizes to the endoplasmic reticulum (ER) in several tissues, including liver, kidney, adrenal glands and macrophages. SOAT1 is involved in cellular cholesterol homeostasis as well as in foam cell formation and the subsequent progression of atherosclerosis. Several SOAT inhibitors have been developed for the treatment of atherosclerosis. SOAT2 (sterol O-acyltransferase-2), also known as ACAT2 (acyl-CoA:cholesterol acyltransferase-2), participates in lipo-protein assembly, catalyzing cholesterol esterification in mammalian cells. SOAT2 is an integral membrane protein that localizes to the endoplasmic reticulum of human intestinal cells. SOAT2 deficiency contributes to severe mental retardation and hypotonus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOAT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOAT1
Gene Accession No. P35610
Gene Alias 8430426K15Rik; Acact; ACACT1; Acat; ACAT1; ACAT-1; acyl coenzyme A:cholesterol acyltransferase 1; acyl-coenzyme A:cholesterol acyltransferase 1; adrenocortical lipid depletion; ald; AW550831; cholesterol acyltransferase 1; hid; SOAT; SOAT1; STAT; sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1; sterol O-acyltransferase 1
Gene Symbols SOAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOAT1. Recombinant protein control fragment (Product #RP-99778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.