Learn More
Invitrogen™ SOAT1 Polyclonal Antibody

Description
Positive test controls include: H460, HeLa, SW480, Mouse stomach, Mouse liver, Rat liver, Rat stomach. The target is usually found in the following locations: Endoplasmic reticulum membrane, Multi-pass membrane protein. Immunogen sequence: MVGEEKMSLR NRLSKSRENP EEDEDQRNPA KESLETPSNG RIDIKQLIAK KIKLTAEAEE LKPFFMKEVG SHFDDFVTNL IEKSASLDNG GCALTTFSVL EGEKNNHRAK DLRAPPEQGK IFIARRSLLD.
Specifications
Specifications
| Antigen | SOAT1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | SOAT1 |
| Gene Accession No. | O70536, P35610, Q61263 |
| Gene Alias | 8430426K15Rik; Acact; ACACT1; Acat; ACAT1; ACAT-1; acyl coenzyme A:cholesterol acyltransferase 1; acyl-coenzyme A:cholesterol acyltransferase 1; adrenocortical lipid depletion; ald; AW550831; cholesterol acyltransferase 1; hid; SOAT; SOAT1; STAT; sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1; sterol O-acyltransferase 1 |
| Gene Symbols | SOAT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.