Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOAT2 Monoclonal Antibody (OTI2E12)
GREENER_CHOICE

Catalog No. PIMA526483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526483 Supplier Invitrogen™ Supplier No. MA526483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOAT2 Monoclonal Antibody for IHC (P)

This gene is a member of a small family of acyl coenzyme A:cholesterol acyltransferases. The gene encodes a membrane-bound enzyme localized in the endoplasmic reticulum that produces intracellular cholesterol esters from long-chain fatty acyl CoA and cholesterol. The cholesterol esters are then stored as cytoplasmic lipid droplets inside the cell. The enzyme is implicated in cholesterol absorption in the intestine and in the assembly and secretion of apolipoprotein B-containing lipoproteins such as very low density lipoprotein (VLDL). Several alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOAT2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOAT2
Gene Accession No. O75908
Gene Alias ACACT2; Acact-2; ACAT2; ACAT-2; acyl Co-A: cholesterol acyltransferase 2; acyl coenzyme A:cholesterol acyltransferase 2; acyl-CoA:cholesterol acyltransferase; acyl-CoA:cholesterol acyltransferase 2; acyl-coenzyme A:cholesterol acyltransferase 2; ARGP2; Cholesterol acyltransferase 2; D15Wsu97e; Soat2; Sterol O-acyltransferase 2
Gene Symbols SOAT2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-120 of SOAT2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8435
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.