Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556091 Supplier Invitrogen™ Supplier No. PA556091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVPIIVPLIP PPFIKPPAED DVSNVQIMCA WCQKVGIKRY SLSMGSEVKS FCSEKCFAAC RRAYFKRNKA RDEDGHAENF PQQH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Implicated in development of the cochlea.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOBP
Gene Accession No. A7XYQ1
Gene Alias jackson circler protein 1; JXC1; MRAMS; sine oculis binding protein homolog; sine oculis-binding protein homolog; SOBP
Gene Symbols SOBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOBP. Recombinant protein control fragment (Product #RP-94688).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.