Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298891G
Description
SOCS-1 Polyclonal specifically detects SOCS-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SOCS-1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SOCS-1. | |
0.1 mL | |
Signal Transduction | |
8651 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction