Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-3 Antibody (OTI3D3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274276
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
SOCS-3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
Mouse | |
24.6 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
OTI3D3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SOCS3 | |
Full length human recombinant protein of human SOCS3 (NP_003946) produced in HEK293T cell. | |
Affinity Purified | |
Signal Transduction | |
9021 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction