Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-3 Antibody (OTI3D3), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274276C
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SOCS-3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SOCS3 | |
Full length recombinant protein of human SOCS-3 (NP_003946) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3D3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
9021.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction