Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-3 Antibody (SO1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001075AF750
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Rat samples. It is validated for Western Blot.Specifications
SOCS-3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SOCS3 | |
Full-length SOCS3 protein. | |
Primary | |
The antibody SO1 reacts with SOCS3, an intracellular cytokine signaling inhibitor. | |
Store at 4°C in the dark. |
Western Blot | |
SO1 | |
Western Blot | |
CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
Mouse | |
0.1 mL | |
9021 | |
Human, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction