Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-4 Antibody (517331) [Janelia Fluor™ 549], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5628I
Description
SOCS-4 Monoclonal antibody specifically detects SOCS-4 in Human,Mouse samples. It is validated for Western BlotSpecifications
SOCS-4 | |
Monoclonal | |
Janelia Fluor 549 | |
CIS4, DKFZp686J1568, SOCS-4, SOCS7, SOCS-7, SSI4, STAT4, suppressor of cytokine signaling 4, Suppressor of cytokine signaling 7SH2 domain containing SOCS box protein | |
E. coli-derived recombinant human SOCS-4, Met1-Cys440, Accession # Q8WXH5 | |
0.1 mL | |
Cell Biology, Signal Transduction | |
122809 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot | |
517331 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction