Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-7/Nck/NAP4 Antibody (23E4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SOCS-7/Nck/NAP4 |
---|---|
Clone | 23E4 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
SOCS-7/Nck/NAP4 Monoclonal antibody specifically detects SOCS-7/Nck/NAP4 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
SOCS-7/Nck/NAP4 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
NAP-4, NAP4SOCS-7, Nck, Ash and phospholipase C binding protein, Nck, Ash and phospholipase C gamma-binding protein, NCKAP4, Nck-associated protein 4, SOCS6, suppressor of cytokine signaling 7 | |
A synthesized peptide derived from Human SOCS-7/Nck/NAP4 [UniProt O14512] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
23E4 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
30837 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title