Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, liquid, OriGene

Rabbit Polyclonal Antibody
Supplier: OriGene TA890038

Description
The Suppressor of cytokine signaling (SOCS) and cytokine-inducible SH2 proteins are a family of intracellular proteins which regulate the immune cell responses to cytokines. SOCS1 acts to suppress dendritic cell (DC) as well as T cell hyperactivation following cytokine signaling by inhibiting JAK tyrosine kinase, a kinase necessary for type I and II cytokine receptors to initiate signaling, by directly binding to the catalytic domain of the kinase. SOCS1 also possesses E3 ubiquitin protein ligase activity that results in the polyubiquitination of its target proteins and subsequent degradation by the proteosome. It is through this method that SOCS1 negatively regulates signaling by Toll-like receptors TLR2 and TLR4 by mediating the degradation of the TLR signaling adaptor protein TIRAP.Specifications
SOCS1 | |
Polyclonal | |
PBS with 50% glycerol and 0.02% sodium azide | |
O15524, O35716, Q9QX78 | |
Socs1 | |
Recombit protein of human SOCS1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SOCS1 | |
CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP-3, TIP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12703, 252971, 8651 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction