Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519813
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519813 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519813 Supplier Invitrogen™ Supplier No. PA519813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat, cow, pig and chimpanzee based on sequence homology.

This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including erythropoietin, GM-CSF, IL10 and interferon (IFN)-gamma. The protein encoded by this gene is found to interact with the cytoplasmic domain of insulin-like growth factor-1 receptor (IGF1R), and thus is thought to be involved in the regulation of IGF1R mediated cell signaling. Knockout studies in mice also suggested a regulatory role of this gene in IGF-1 related growth control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene SOCS2
Gene Accession No. O14508, O35717
Gene Alias 8030460M17; AI527257; AW108012; CIS2; CIS-2; Cish2; cytokine inducible SH2-containing protein 2; cytokine-inducible SH2 protein 2; D130043N08Rik; hg; JAB; Socs2; SOCS-2; SSI2; SSI-2; STAT induced STAT inhibitor-2; STATI2; STAT-induced STAT inhibitor 2; STAT-induced STAT inhibitor-2; Suppressor of cytokine signaling 2; suppressor of cytokine signaling-2
Gene Symbols SOCS2
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human SOCS2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216233, 8835
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.