Learn More
SOCS4 Mouse anti-Human, Clone: OTI5H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811167

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
SOCS4 | |
Monoclonal | |
Unconjugated | |
SOCS4 | |
SOCS7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
122809 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WXH5 | |
SOCS4 | |
Full length human recombit protein of human SOCS4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.