Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOCS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521599
Description
Recommended positive controls: HeLaS3. Predicted reactivity: Pig (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| SOCS4 | |
| Polyclonal | |
| Unconjugated | |
| SOCS4 | |
| 3110032M18Rik; A730004F22Rik; AI427843; AU015727; AU015859; SH2 domain containing SOCS box protein; SOCS4; SOCS-4; SOCS7; SOCS-7; suppressor of cytokine signaling 4; Suppressor of cytokine signaling 7; suppressor of cytokine signalling 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122809 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WXH5 | |
| SOCS4 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SOCS4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction