Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566116 Supplier Invitrogen™ Supplier No. PA566116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYVWSGKKLSW SKKSESYSDA ETVNGIEKTE VSLRNQERKH SCSSIELDLD HSCGHRFLGR SLKQKLQDAV GQCFPIKNCS SRHSS Highest antigen sequence identity to the following orthologs - mouse 80%, rat 78%.

The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOCS4
Gene Accession No. Q8WXH5
Gene Alias 3110032M18Rik; A730004F22Rik; AI427843; AU015727; AU015859; SH2 domain containing SOCS box protein; SOCS4; SOCS-4; SOCS7; SOCS-7; suppressor of cytokine signaling 4; Suppressor of cytokine signaling 7; suppressor of cytokine signalling 4
Gene Symbols SOCS4
Host Species Rabbit
Immunogen Recombinant Human SOCS4. Recombinant protein control fragment (Product #RP-106803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.