Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582737 Supplier Invitrogen™ Supplier No. PA582737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLIKQHTAPV SPHSTFFDTF DPSLVSTEDE EDRLRERRRL SIEEGVDPPP NAQIHTFEAT AQVNPLYKLG PKLAPGMTEI SGDSSAIPQA NCDSEEDTTT LCLQSRRQKQ RQISGDSHTH VSRQGAWKVH TQI Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 95%.

Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOCS5
Gene Accession No. O75159
Gene Alias 1810018L08Rik; CIS6; CIS-6; CISH5; CISH6; cytokine inducible SH2-containing protein 5; cytokine-inducible SH2 protein 6; Cytokine-inducible SH2-containing protein 5; KIAA0671; mKIAA0671; RGD1564914; Socs5; SOCS-5; Suppressor of cytokine signaling 5
Gene Symbols SOCS5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOCS5. Recombinant protein control fragment (Product #RP-102073).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.