Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS6 Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502281654
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1654 Supplier Bioss Supplier No. BS10685RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOCS6 Polyclonal antibody specifically detects SOCS6 in Chicken, Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SOCS6
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene SOCS6
Gene Accession No. O14544, Q9JLY0
Gene Alias 1500012M23Rik; 5830401B18Rik; AI447482; Cis4; CIS-4; Cish4; cytokine inducible SH2-containing protein 4; cytokine inducible SH2-containing protein CIS4; cytokine-inducible SH2 protein 4; HSPC060; SH2 domain containing SOCS box protein SOCS6; SOCS4; SOCS-4; Socs6; SOCS-6; SSI4; STAI4; STAT induced STAT inhibitor-4; STATI4; Suppressor of cytokine signaling 4; suppressor of cytokine signaling 6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SOCS6 (21-120).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 421020, 54607, 9306
Target Species Chicken, Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.