Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, 1M Buffer Solution, pH 8.5, Thermo Scientific Chemicals PROMO

Catalog No. J60408AKPM
Click to view available options
:
250 mL

Applications

  • Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source.
  • This solution prepared in water is used as injection in Pharmaceutical industry. It is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.
  • It is also used as a buffer.
  • Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility

  • Soluble in water

Notes

  • Store in cool place.
  • Keep container tightly closed in a dry and well-ventilated place.
  • Keep away fom strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01 g/mol
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in Water
Chemical Name or Material Sodium bicarbonate buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.