Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium Phosphate, 0.5M Buffer Solution, pH 8.0 PROMO

Catalog No. J60825APPM
Click to view available options
:
500 mL

Applications

  • Sodium phosphate buffer is used routinely in the life sciences.
  • It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM.
  • Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility

  • Soluble in water (121 mg/ml at 20°C).

Notes

  • Store in cool place.
  • Keep container tightly closed in a dry and well-ventilated place.
  • Keep away from strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
MDL Number MFCD00146206
Solubility Information Soluble in Water (121mg/ml at 20°C).

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.