Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD1/Cu-Zn SOD Antibody (106), Alexa Fluor™ 750

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289969AF750
Description
SOD1/Cu-Zn SOD Monoclonal specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Flow Cytometry.Specifications
SOD1/Cu-Zn SOD | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
106 | |
Flow Cytometry | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD1/Cu-Zn SOD (NP_000445.1; Ala2-Gln154). | |
0.1 mL | |
Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
6647 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction