Learn More
Invitrogen™ SOD1 Polyclonal Antibody
Description
SOD1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | SOD1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SOD1 |
| Gene Accession No. | P09212 |
| Gene Alias | ALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodb |
| Gene Symbols | SOD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.