Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOD1 Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142219
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142219 Supplier Invitrogen™ Supplier No. PA5142219
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SOD1
Gene Accession No. P00441, P07632, P08228
Gene Alias ALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodb
Gene Symbols SOD1
Host Species Goat
Immunogen Peptide with sequence C-SRKHGGPKDEERH (aa 69-81).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20655, 24786, 6647
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.