Learn More
SOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human SOD1.
Supplier: Abnova Corporation PAB29364
Description
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq]
Sequence: PVQGIINFEQKESNGPVKVWGSIKGLTEGLHGFHVHEFGDNTAGCTSAGPHFNPLSRKHGGPKDEERHVGDLGNVTADKDGVADVSIEDSVISLSGDHCIIGRTLVVHEKADDLGKGGNEESTKTGNAGSRLACGSpecifications
SOD1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human SOD1. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
SOD1 | |
Recombinant protein corresponding to human SOD1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:2500-1:5000) Immunofluorescence (1-4 ug/mL) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
ALS/ALS1/IPOA/SOD/homodimer | |
Rabbit | |
Antigen affinity purification | |
RUO | |
6647 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.