Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOD1 Recombinant Rabbit Monoclonal Antibody (101)
GREENER_CHOICE

Catalog No. PIMA529576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529576 Supplier Invitrogen™ Supplier No. MA529576
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Superoxide Dismutase.

SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD1
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 101
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SOD1
Gene Accession No. P00441
Gene Alias ALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodb
Gene Symbols SOD1
Host Species Rabbit
Immunogen Recombinant Human SOD1 protein (Ala2-Gln154).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.