Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD2/Mn-SOD, ac Lys68 Antibody (23G5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SOD2/Mn-SOD |
---|---|
Clone | 23G5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
SOD2/Mn-SOD | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
A synthesized peptide derived from Human SOD2/Mn-SOD [UniProt P04179] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
23G5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6648 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title