Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD2/Mn-SOD Antibody (012), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290139APC
Description
SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SOD2/Mn-SOD | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
012 | |
Immunohistochemistry-Paraffin | |
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD2/Mn-SOD (Uniprot#: P04179-1; Lys25-Lys222). | |
0.1 mL | |
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
6648 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction