Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD2/Mn-SOD Antibody (10F6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32630850UL
Description
SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
SOD2/Mn-SOD | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
10F6 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
A synthesized peptide derived from Human SOD2/Mn-SOD [UniProt P04179] | |
50 μL | |
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
6648 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction