Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD2/Mn-SOD Antibody, Biotin, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB200165B
Description
SOD2/Mn-SOD Polyclonal antibody specifically detects SOD2/Mn-SOD in Rat samples. It is validated for Western BlotSpecifications
SOD2/Mn-SOD | |
Polyclonal | |
Western Blot | |
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
A synthetic peptide between amino acids 175-222 of human manganese superoxide dismutase encoded within exon 5 (LocusLink ID 6648). | |
0.1 mL | |
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
6648 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction