Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2 Mouse anti-Bovine, Human, Clone: 9E2BD2, Abnova™

Catalog No. 89035565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-565 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-565 Supplier Abnova Corporation Supplier No. MAB14167

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against bovine and pig SOD2.

  • High specificity
  • High reactivity

Specifications

Antigen SOD2
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9E2BD2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against bovine and pig SOD2.
Dilution ELISA Immunocytochemistry (2 ug/mL) Immunofluorescence (2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the e
Formulation In HEPES (0.02% sodium azide).
Gene Alias MGC128371
Gene Symbols SOD2
Host Species Mouse
Immunogen Purified proteins from bovine liver, bovine heart, porcine heart.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281496/100154319
Target Species Bovine, Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.