Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2 Mouse anti-Human, Clone: 37CT127.5.11.6, Abnova™

Catalog No. 89934946 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-946 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-946 Supplier Abnova Corporation Supplier No. MAB12323
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human SOD2.

This gene is a member of the iron/manganese superoxide dismutase family. It encodes a mitochondrial protein that forms a homotetramer and binds one manganese ion per subunit. This protein binds to the superoxide byproducts of oxidative phosphorylation and converts them to hydrogen peroxide and diatomic oxygen. Mutations in this gene have been associated with idiopathic cardiomyopathy (IDC), premature aging, sporadic motor neuron disease, and cancer. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Specifications

Antigen SOD2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 37CT127.5.11.6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human SOD2.
Dilution Immunohistochemistry (1:10-1:50) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias IPO-B/MNSOD/Mn-SOD
Gene Symbols SOD2
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human SOD2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6648
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.