Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD3/EC-SOD Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | SOD3/EC-SOD |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SOD3/EC-SOD Polyclonal specifically detects SOD3/EC-SOD in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SOD3/EC-SOD | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse | |
EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
SOD3 | |
Primary | |
Specific for SOD3. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Sterilized water with No Preservative | |
6649 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title