Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOD3 Monoclonal Antibody (C1)

Catalog No. PIMA543570
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543570 20 μL
PIMA541679 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543570 Supplier Invitrogen™ Supplier No. MA543570
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOD3 Monoclonal Antibody for Western Blot

This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD3
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SOD3
Gene Accession No. P08294
Gene Alias AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175
Gene Symbols SOD3
Host Species Mouse
Immunogen Recombinant protein Superoxide Dismutase 3, Extracellular. The antigen corresponds to amino acid range 19-240 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.