Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580051
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580051 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580051 Supplier Invitrogen™ Supplier No. PA580051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Human Placenta Tissue, A549 Cell, MM231 Cell, MCF-7 Cell. IHC: human mammary cancer tissue IHC-F: Human Placenta Tissue. ICC/IF: HELA cell.

This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SOD3
Gene Accession No. P08294
Gene Alias AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175
Gene Symbols SOD3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human SOD3 (41-56aa KVTEIWQEVMQRRDDD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6649
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.