Learn More
SODD/BAG4 Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP176897SS
Catalog No. NBP176897SS
Only null left
Add to Cart
Description
Description
SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SODD/BAG4
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4
Rabbit
60 kDa
0.025 mg
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/ml
Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL
AF111116
BAG4
Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human SODD. The immunogen is located within the first 50 amino acids of SODD. Amino Acid Squence: SALRRSGYGPSDGPS
Ion exchange chromatography
Apoptosis, Cancer, Tumor Suppressors
9530
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.