Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Naphthalenesulfonate 98.0+%, TCI America™
SDP

Catalog No. N0015100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0015100G 100 g
N001525G 25 g
N0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0015100G Supplier TCI America Supplier No. N0015100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130-14-3
Molecular Formula C10H7NaO3S
Molecular Weight (g/mol) 230.213
MDL Number MFCD00064964
InChI Key HIEHAIZHJZLEPQ-UHFFFAOYSA-M
Synonym sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn
PubChem CID 23661867
IUPAC Name sodium;naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+]

Specifications

Color White
pH 5.98 (5% w/v H2O soln. 20°C)
Quantity 100 g
Formula Weight 230.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthalenesulfonate
EINECSNumber (4)-0466
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.