Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Propanesulfonate 98.0+%, TCI America™
SDP

Catalog No. P034725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P034725G 25 g
P03475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P034725G Supplier TCI America Supplier No. P034725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14533-63-2
Molecular Formula C3H7NaO3S
Molecular Weight (g/mol) 146.136
MDL Number MFCD00062546
InChI Key NPAWNPCNZAPTKA-UHFFFAOYSA-M
Synonym sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt
PubChem CID 4319363
IUPAC Name sodium;propane-1-sulfonate
SMILES CCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 146.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Propanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.