Learn More
Sodium 2-ethylhexanoate, 97%
Description
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 1000 g |
Solubility Information | Soluble in water. |
Formula Weight | 166.2 |
Percent Purity | 97% |
Chemical Name or Material | Sodium 2-ethylhexanoate |
Safety and Handling
Hazard Category | H361 |
Hazard Statement | GHS H Statement H361 Suspected of damaging fertility or the unborn child. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 243-283-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.