Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium 2-Naphthalenesulfonate 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | White |
Quantity | 25 g |
Formula Weight | 230.21 |
Percent Purity | ≥98.0% (HPLC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Sodium 2-Naphthalenesulfonate |
Safety and Handling
EINECSNumber | (4)-0466 |
RTECSNumber | QK3678000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction