Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-Bromopropanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B1707100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1707100G 100 g
B170725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1707100G Supplier TCI America Supplier No. B1707100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55788-44-8
Molecular Formula C3H7BrNaO3S
Molecular Weight (g/mol) 226.04
MDL Number MFCD00024999
InChI Key ONPNLMMBTJZJRQ-UHFFFAOYSA-N
Synonym sodium 3-bromopropanesulfonate, sodium 3-bromopropane-1-sulfonate, 3-bromopropanesulfonic acid sodium salt, acmc-209lpd, sodium 3-bromo-1-propanesulfonate, sodium 3-bromo-propane-1-sulphonate, 3-bromopropane sulfonic acid sodium salt, 3-bromo-1-propanesulfonic acid sodium salt, 1-propanesulfonic acid, 3-bromo-, sodium salt
PubChem CID 23679056
IUPAC Name 3-bromopropane-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)CCCBr

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 225.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Bromopropanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.