Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 3-mercapto-1-propanesulfonate

Catalog No. 25168225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-168-225G 25 g
25-168-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-168-225G Supplier Sigma Aldrich Supplier No. 25168225G

Due to product restrictions, please to purchase or view availability for this product.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 3-mercapto-1-propanesulfonate

Specifications

CAS 17636-10-1
Melting Point ∼220°C (lit.)
Molecular Formula C3H7NaO3S2
Linear Formula HSCH2CH2CH2SO3Na
MDL Number MFCD00043148
Quantity 25 g
Synonym 3-Mercapto-1-propanesulfonic acid sodium salt; MPS
Molecular Weight (g/mol) 178.21
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.