Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Acetate, 20% (w/w), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
CAS | 7732-18-5 |
Molecular Weight (g/mol) | 136.08 g/mol |
Percent Purity | 20% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Methods | APHA for Iron |
pH | 7.5 to 9.2 |
Quantity | 4 L |
Chemical Name or Material | Sodium Acetate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction