Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate/Acetic Acid Buffer, Reagents

Catalog No. 01333766
Click to view available options
:
Quantity: 20 L

Catalog No. 01-333-766

Supplier: Reagents Holdings Llc CS12370020E7

Only null left
Add to Cart
Add to Cart

Sodium Acetate/Acetic Acid Buffer, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Acetate/Acetic Acid Buffer
CAS 7732-18-5, 64-19-7, 6131-90-4
Color Colorless
Synonym Ethanoic Acid/Sodium acetate
Grade Reagent
Quantity 20 L
Packaging HDPE Winpak
Physical Form Liquid
Specific Gravity 1.0749
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.