Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number AA1100506
Click to view available options
Quantity:
5 g
25 g
100 g
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O

Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 324°C
Color Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.997% (metals basis)
Beilstein 3595639
Solubility Information Very soluble in water. Soluble in alcohol.
Formula Weight 82.03
Odor Nearly odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 1.528
Chemical Name or Material Sodium acetate, anhydrous
Show More Show Less
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4300010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit