Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Anhydrous, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS022525A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS022525A 2.5 kg
RDCS0225500 500 g
RDCS022512F 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RDCS022525A Supplier Ricca Chemical Company Supplier No. RDCS02252.5A5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

pH 7.0-9.2
CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 2.5 kg
Percent Purity min 99%
Grade ACS
Loss on Drying max. 1.0%
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Acetate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.